Learn More
Medchemexpress LLC HY-N2332A 5mg Medchemexpress, Methyllycaconitine citrate CAS:112825-05-5 Purity:>98%
SDP
Catalog No. 501933912
Encompass_Preferred
Medchemexpress, HY-N2332A 5mg Methyllycaconitine citrate CAS:112825-05-5 Methyllycaconitine citrate is a specific antagonist of α7 neuronal nicotinic acetylcholine receptor ( α7nAChR ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3912 Supplier Medchemexpress LLC Supplier No. HYN2332A5MG
Medchemexpress, HY-N2332A 5mg Methyllycaconitine citrate CAS:112825-05-5 Methyllycaconitine citrate is a specific antagonist of α7 neuronal nicotinic acetylcholine receptor ( α7nAChR ). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.