Learn More
Abcam Methyllycaconitine citrate (MLA), alpha7 Nicotinic receptor antagonist, 25MG
Catalog No. 502584708
Encompass_Preferred
MW 874.9 Da, Purity >98%. Potent, selective antagonist at α7-containing nicotinic receptors. Also available in simple stock solutions (ab146684) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4708 Supplier Abcam Supplier No. AB12007225MG
MW 874.9 Da, Purity >98%. Potent, selective antagonist at α7-containing nicotinic receptors. Also available in simple stock solutions (ab146684) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement