Learn More
Abcam Methyllycaconitine citrate (MLA) (mM/ml), alpha7 Nicotinic receptor antagonist, 10MM_ML
Catalog No. 502704528
Encompass_Preferred
MW 874.92 g/mol, Purity >98%. Potent, selective antagonist at α7-containing nicotinic receptors. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4528 Supplier Abcam Supplier No. AB14668410MM_ML
MW 874.92 g/mol, Purity >98%. Potent, selective antagonist at α7-containing nicotinic receptors. Achieve your results faster with highly validated, pure and trusted compounds.
The product is subject to the following: Abcam Restricted Use Statement