Learn More
Abcam Methyllycaconitine citrate (MLA), alpha7 Nicotinic receptor antagonist, 5MG
Catalog No. 502584707
Encompass_Preferred
MW 874.9 Da, Purity >98%. Potent, selective antagonist at α7-containing nicotinic receptors. Also available in simple stock solutions (ab146684) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-258-4707 Supplier Abcam Supplier No. AB1200725MG
MW 874.9 Da, Purity >98%. Potent, selective antagonist at α7-containing nicotinic receptors. Also available in simple stock solutions (ab146684) - add 1 ml of water to get an exact, ready-to-use concentration.
The product is subject to the following: Abcam Restricted Use Statement