Learn More
Strem, An Ascensus Company CAS 75-16-1. 1mole. Methylmagnesium bromide 3M in ether. MFCD00000041
SDP
Catalog No. 502110870
Encompass
CAS 75-16-1. 1mole. Methylmagnesium bromide 3M in ether. MFCD00000041. Molecular Weight 119.26. Molecular Formula CH3MgBr. Color/form liq. Strem 93-1265. http//www.strem.com/catalog/v/93-1265/
Catalog No. 50-211-0870 Supplier Strem, An Ascensus Company Supplier No. 9312651MOL
CAS 75-16-1. 1mole. Methylmagnesium bromide 3M in ether. MFCD00000041. Molecular Weight 119.26. Molecular Formula CH3MgBr. Color/form liq. Strem 93-1265. http//www.strem.com/catalog/v/93-1265/