Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylmagnesium bromide, 3M in ether, packaged under Argon in resealable ChemSeal™ bottles

Catalog No. AA41252AP
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA41252AP 500 mL
AA41252AE 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA41252AP Supplier Thermo Scientific Chemicals Supplier No. 041252AP

missing translation for 'validMainframeMsg'

CAS: 75-16-1 | CH3BrMg | 119.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Methylmagnesium bromide, 3M in ether has the wide range of applications in organic synthesis. Alkylation of aldehydes and ketones is the most common applications of Grignard reagent. It is a strong nucleophile that can form new carbon-carbon bonds. Also like organolithium compounds, it is useful to form carbon-heteroatom bonds.

Solubility
Soluble in ether, toluene and tetrahydrfuran. Insoluble in hydrocarbons.

Notes
It is highly flammable and reacts with moisture or water to form methane gas, so it should be stored in tightly closed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 75-16-1
Molecular Formula CH3BrMg
Molecular Weight (g/mol) 119.24
MDL Number MFCD00000041
InChI Key AVFUHBJCUUTGCD-UHFFFAOYSA-M
PubChem CID 6349
SMILES C[Mg]Br

Specifications

Color Brown
Physical Form Liquid
Quantity 500 mL
Linear Formula CH3MgBr
UN Number UN1928
Flash Point -40°C (-40°F)
Synonym methyl magnesium bromide, grignard reagent
IUPAC Name bromo(methyl)magnesium
Formula Weight 119.26
Concentration or Composition (by Analyte or Components) 3M in ether
Odor Ether-like
Packaging ChemSeal™ bottles
Sensitivity Air and moisture sensitive
Density 1.035 g/mL
Chemical Name or Material Methylmagnesium bromide
Show More Show Less
Hazard Category H224-H260-H302-H314-H318-H335-H336
Hazard Statement GHS H Statement
H224-H261-H314-H318-H302-H336
Extremely flammable liquid and vapor.
In contact with water releases flammable gas.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
May cause drowsiness or dizziness.
Precautionary Statement P210-P231+P232-P233-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P331-P363-P372-P374-P380-P501c
DOTInformation Transport Hazard Class: 4.3; Packaging Group: I; Proper Shipping Name: METHYL MAGNESIUM BROMIDE, IN ETHYL ETHER
EINECSNumber 200-844-1
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.