Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylmagnesium Bromide (ca. 30% in 2-Methyltetrahydrofuran, ca. 3mol/L), TCI America™
SDP

Catalog No. M2237250G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M2237250G 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M2237250G Supplier TCI America Supplier No. M2237250G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 75-16-1
Molecular Formula CH3BrMg
Molecular Weight (g/mol) 119.24
MDL Number MFCD00000041
InChI Key AVFUHBJCUUTGCD-UHFFFAOYSA-M
Synonym methylmagnesium bromide, methyl magnesium bromide, magnesium, bromomethyl, bromo methyl magnesium, methylmagnesium bromide solution, 3.0 m in diethyl ether, unii-22cw9773df, memgbr, methymagnesiumbromide, ch3mgbr, methylmagnesiumbromide
PubChem CID 6349
IUPAC Name bromo(methyl)magnesium
SMILES C[Mg]Br

Specifications

Quantity 250 g
UN Number 3399
Formula Weight 119.24
Chemical Name or Material Methylmagnesium Bromide (ca. 30% in 2-Methyltetrahydrofuran, ca. 3mol/L)
EINECSNumber (2)-3468
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.