Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich Methylmagnesium Bromide Solution

Catalog No. 1898988L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mL
100 mL
4 x 25 mL
800 mL
4 x 100 mL
8 L
18 L
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
18-989-88L 8 L
50-308-1448 4 x 25 mL
50-308-1446 100 mL
18-989-850ML 50 mL
18-989-8800ML 800 mL
18-989-818LKL 18 L
50-308-1447 4 x 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Danger
Catalog No. 18-989-88L Supplier Sigma Aldrich Supplier No. 1898988L
Add to Cart
Add to Cart
Danger

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

Methylmagnesium bromide solution (3M in diethyl ether) be used to synthesize:3(4R)-{3(2R,3S)-[4'sa-(1'sa-triphenylmethyl)imidazole]-2-bromo-1-oxobutyl)-4-phenyl2-oxazolidinoneBenzyl 2-(2-oxopropyl)piperidine-1-carboxylate(R, R) 1-[1-(1-phenylethyl)pyrrolidin-2-yl]propan-2-one)Sure/Seal is a trademark of Sigma-Aldrich Co. LLC

Specifications

CAS 75-16-1
Density 1.035 g/mL at 25°C
Molecular Formula CH3BrMg
Linear Formula CH3MgBr
MDL Number MFCD00000041
Quantity 8 L
Molecular Weight (g/mol) 119.24 g/mol
Biological Activity Central Nervous System
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.