Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Sigma Aldrich Methylmagnesium Chloride Solution
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Methylmagnesium chloride solution (3 M in THF) undergoes 1,3-addition with dialdose derived nitrones to form N-benzylhydroxylamines. The reductive cleavage of the N-O bond of these N-benzylhydroxylamines results in N-benzylaminosugars, which are key moieties of bioactive compounds. It can also react with Garner type aldehydes to form the corresponding secondary alcohols.Sure/Seal is a trademark of Sigma-Aldrich Co. LLC
Specifications
Specifications
| CAS | 676-58-4 |
| Density | 1.013 g/mL at 25°C |
| Molecular Formula | CH3ClMg |
| Linear Formula | CH3MgCl |
| Quantity | 800 mL |
| MDL Number | MFCD00000468 |
| Molecular Weight (g/mol) | 74.79 g/mol |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?