Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylmaleic Hydrazide 98.0+%, TCI America™
SDP

Catalog No. M218225G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M218225G 25 g
M21825G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M218225G Supplier TCI America Supplier No. M218225G

Chemical Identifiers

CAS 5754-18-7
Molecular Formula C5H6N2O2
Molecular Weight (g/mol) 126.12
MDL Number MFCD00067746,MFCD00023867
InChI Key QAVYOWFNXMHVEL-UHFFFAOYSA-N
Synonym 4-methylpyridazine-3,6-diol, 3,6-dihydroxy-4-methylpyridazine, 1,2-dihydro-4-methyl-3,6-pyridazinedione, 4-methyl-3,6-pyridazinediol, 3,6-pyridazinedione, 1,2-dihydro-4-methyl, 3, 1,2-dihydro-4-methyl, 2-butenedioic acid, cyclic hydrazide, z, acmc-1aqja, 4-methylmaleic hydrazide, methylmaleic acid hydrazide
PubChem CID 79826
IUPAC Name 4-methyl-1,2,3,6-tetrahydropyridazine-3,6-dione
SMILES CC1=CC(=O)NNC1=O

Specifications

Melting Point 287°C
Color White-Yellow
Quantity 25 g
Formula Weight 126.12
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methylmaleic Hydrazide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.