Learn More
Medchemexpress LLC Methylmalonic acid | 516-05-2 | MFCD00002656 | 99.2% | 118.09 g/mol | C4H6O4 | 1 G
SDP
Catalog No. 5000245114
Encompass_Preferred
Methylmalonic acid is an organic dicarboxylic acid used as an analytical standard and a biomarker for vitamin B12 deficiency. It is used in analytical assays, calibration, and method development.
  • Analytical standard for assay calibration and reference measurements.
  • Molecular formula C4H6O4; molecular weight 118.09 g/mol.
  • High purity (≈99.2%) for reliable quantitation.
  • Suitable for research and analytical applications, soluble in water and DMSO.
Catalog No. 50-002-45114 Supplier Medchemexpress LLC Supplier No. HY1033951G
Add to Cart
Add to Cart
Methylmalonic acid is an organic dicarboxylic acid used as an analytical standard and a biomarker for vitamin B12 deficiency. It is used in analytical assays, calibration, and method development.
  • Analytical standard for assay calibration and reference measurements.
  • Molecular formula C4H6O4; molecular weight 118.09 g/mol.
  • High purity (≈99.2%) for reliable quantitation.
  • Suitable for research and analytical applications, soluble in water and DMSO.