Learn More
Medchemexpress LLC Methylmalonic acid | 516-05-2 | MFCD00002656 | 99.2% | 118.09 g/mol | C4H6O4 | 50 MG
SDP
Catalog No. 5000244557
Encompass_Preferred
Methylmalonic acid (methylpropanedioic acid) analytical standard intended for research and analytical use. Supplied as a high-purity reference material (listed purity 99.19%), it is suitable for calibration, method validation, and as a biomarker reference in studies of vitamin B12 status.
  • Analytical standard for research and analytical applications.
  • High purity suitable for calibration and method validation.
  • Used as a biomarker reference related to vitamin B12 deficiency.
  • Available in small weighed quantities for precise dosing, including 50 mg packs.
  • Synonyms include methylpropanedioic acid and methylmalonate.
Catalog No. 50-002-44557 Supplier Medchemexpress LLC Supplier No. HY103395R50MG
Add to Cart
Add to Cart
Methylmalonic acid (methylpropanedioic acid) analytical standard intended for research and analytical use. Supplied as a high-purity reference material (listed purity 99.19%), it is suitable for calibration, method validation, and as a biomarker reference in studies of vitamin B12 status.
  • Analytical standard for research and analytical applications.
  • High purity suitable for calibration and method validation.
  • Used as a biomarker reference related to vitamin B12 deficiency.
  • Available in small weighed quantities for precise dosing, including 50 mg packs.
  • Synonyms include methylpropanedioic acid and methylmalonate.