Learn More
Cayman Chemical Methylmalonic Acid, Purity: ≥95%, CAS: 516-05-2, 5 g
SDP
Catalog No. 502972839
Encompass_Preferred
A dicarboxylic acid and byproduct of propionate catabolism; impairs mitochondrial respiration, increases glucose consumption, and downregulates the transcription of ENO2 and SYP in neuronally differentiated SH-SY5Y cells; intrastriatal adminsitration induces neuronal loss in rats at 100 mg/ml; accumulates in the tissues and body fluids of patients with methylmalonic acidemia
Catalog No. 50-297-2839 Supplier Cayman Chemical Supplier No. 148855 G
A dicarboxylic acid and byproduct of propionate catabolism; impairs mitochondrial respiration, increases glucose consumption, and downregulates the transcription of ENO2 and SYP in neuronally differentiated SH-SY5Y cells; intrastriatal adminsitration induces neuronal loss in rats at 100 mg/ml; accumulates in the tissues and body fluids of patients with methylmalonic acidemia