Learn More
Cayman Chemical MethylmalonIc AcId-d3 10mg
SDP
Catalog No. 502913464
Encompass_Preferred
An internal standard for the quantification of methylmalonic acid by GC- or LC-MS
Catalog No. 50-291-3464 Supplier Cayman Chemical Supplier No. 3523210 MG
An internal standard for the quantification of methylmalonic acid by GC- or LC-MS