Learn More
Cayman Chemical Methylmalonic Acid-d3, Purity: ≥99% deuterated forms (d1-d3), CAS: 42522-59-8, 25 mg
SDP
Catalog No. 502913406
Encompass_Preferred
An internal standard for the quantification of methylmalonic acid by GC- or LC-MS
Catalog No. 50-291-3406 Supplier Cayman Chemical Supplier No. 3523225 MG
An internal standard for the quantification of methylmalonic acid by GC- or LC-MS