Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methylmalonic acid-d3 | 42522-59-8 | MFCD00144783 | >98.0% | 121.1 g/mol | C4H3D3O4 | 10 MG
SDP

Catalog No. 5000244578
Encompass_Preferred

Methylmalonic acid-d3 is the deuterium-labeled analogue of methylmalonic acid used as an internal standard for quantitative GC- or LC-MS analyses. It is intended for research and analytical applications and is not for human or clinical use.

  • Deuterium-labeled (d3) methyl group for use as an internal standard in mass spectrometry.
  • High deuterium incorporation: ≥99% (d1-d3) per certificate of analysis.
  • Chemical purity ≥98% (methylmalonic acid basis).
  • Molecular formula C4H3D3O4; molecular weight 121.1 g/mol.
  • Supplied as a solid; commonly available in 5 MG, 10 MG, 25 MG, and 50 MG pack sizes.
  • Intended for accurate quantification of methylmalonic acid by GC- or LC-MS.

Catalog No. 50-002-44578 Supplier Medchemexpress LLC Supplier No. HY103395S10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylmalonic acid-d3 is the deuterium-labeled analogue of methylmalonic acid used as an internal standard for quantitative GC- or LC-MS analyses. It is intended for research and analytical applications and is not for human or clinical use.

  • Deuterium-labeled (d3) methyl group for use as an internal standard in mass spectrometry.
  • High deuterium incorporation: ≥99% (d1-d3) per certificate of analysis.
  • Chemical purity ≥98% (methylmalonic acid basis).
  • Molecular formula C4H3D3O4; molecular weight 121.1 g/mol.
  • Supplied as a solid; commonly available in 5 MG, 10 MG, 25 MG, and 50 MG pack sizes.
  • Intended for accurate quantification of methylmalonic acid by GC- or LC-MS.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.