Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methylmalonic acid-d3 | 42522-59-8 | MFCD00144783 | 98.9% | 121.11 | C4H3D3O4 | 25 MG
SDP

Catalog No. 5000244653
Encompass_Preferred

Methylmalonic acid-d3 is the deuterium (d3) labeled analogue of methylmalonic acid for research and analytical use. It is used as an internal standard and tracer for quantitative analysis by GC-MS and LC-MS, and in metabolic studies related to vitamin B12 status.

  • Deuterium-labeled (d3) internal standard for GC-MS and LC-MS.
  • High purity (98.9%) suitable for analytical applications.
  • Molecular formula C4H3D3O4; molecular weight 121.11.
  • White to off-white solid supplied in small quantities for standards and calibration.
  • Recommended storage: powder -20°C for long-term stability.

Catalog No. 50-002-44653 Supplier Medchemexpress LLC Supplier No. HY103395S25MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylmalonic acid-d3 is the deuterium (d3) labeled analogue of methylmalonic acid for research and analytical use. It is used as an internal standard and tracer for quantitative analysis by GC-MS and LC-MS, and in metabolic studies related to vitamin B12 status.

  • Deuterium-labeled (d3) internal standard for GC-MS and LC-MS.
  • High purity (98.9%) suitable for analytical applications.
  • Molecular formula C4H3D3O4; molecular weight 121.11.
  • White to off-white solid supplied in small quantities for standards and calibration.
  • Recommended storage: powder -20°C for long-term stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.