Learn More
Cayman Chemical MethylmalonIc AcId-d3 5mg
SDP
Catalog No. 502909335
Encompass_Preferred
An internal standard for the quantification of methylmalonic acid by GC- or LC-MS
Catalog No. 50-290-9335 Supplier Cayman Chemical Supplier No. 352325 MG
An internal standard for the quantification of methylmalonic acid by GC- or LC-MS