Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylmercury(II) hydroxide, 1M in H{2}O

Catalog No. AA13395AD
Change view
Click to view available options
Quantity:
10 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA13395AD 50 mL
AA13395AB 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA13395AD Supplier Thermo Scientific Chemicals Supplier No. 013395AD
Add to Cart

missing translation for 'validMainframeMsg'

As a defoliant and epoxidation catalyst, as denaturing agent | CAS: 1184-57-2

Methyl mercury(II) hydroxide is used as defoliant and epoxidation catalyst, as denaturing agent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1184-57-2
MDL Number MFCD00013924

Specifications

Color Colorless
Physical Form Liquid
Quantity 50 mL
Linear Formula CH3HgOH
UN Number UN2024
Solubility Information Fully miscible in water.
Formula Weight 232.63
Concentration or Composition (by Analyte or Components) 1M in H2O
Chemical Name or Material Methylmercury(II) hydroxide
Hazard Category H300+H310+H330-H373
Hazard Statement GHS H Statement
H300-H311-H331-H373
Fatal if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P260-P262-P264b-P270-P271-P280-P284-P301+P310-P302+P350-P304+P340-P310-P314-P330-P361-P363-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: MERCURY COMPOUNDS, LIQUID, N.O.S.
EINECSNumber 214-667-2
RTECSNumber OW4900000
TSCA No
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.