Learn More
Crescent Chemical Co Inc METHYLNADIC ANHYDRIDE PRACT.
SDP
Supplier:  Crescent Chemical Co Inc SE29452.01
Encompass_Preferred
Methylnadic anhydride pract. Hardener component for polyester and epoxy resins, for example EPON embedding. Size - 100ML Storage Conditions - +15 °C TO +30 °C Catalog Number - 29452.01 CAS 25134-21-8
Catalog No. 50-247-861