Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methylnissolin-3-O-glucoside | 94367-42-7 | 5 MG
SDP

Catalog No. 5000420813
Encompass_Preferred

Methylnissolin-3-O-glucoside is a flavonoid derived from the roots of Astragalus membranaceus, known for its anti-inflammatory effects.

  • Purity: 99.91%
  • Molecular weight: 462.45
  • Chemical formula: C23H26O10
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Flavonoids
  • Store at 4°C, protect from light
  • Solubility in DMSO: 100 mg/mL

Catalog No. 50-004-20813 Supplier Medchemexpress LLC Supplier No. HYN24735MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylnissolin-3-O-glucoside is a flavonoid derived from the roots of Astragalus membranaceus, known for its anti-inflammatory effects.

  • Purity: 99.91%
  • Molecular weight: 462.45
  • Chemical formula: C23H26O10
  • Appearance: Solid
  • Color: White to off-white
  • Structure classification: Flavonoids
  • Store at 4°C, protect from light
  • Solubility in DMSO: 100 mg/mL

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.