Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methylnissolin-3-O-glucoside | 94367-42-7 | 99.9% | 462.45 g/mol | C23H26O10 | 10 MG
SDP

Catalog No. 5000289471
Encompass_Preferred

Methylnissolin-3-O-glucoside is a flavonoid glycoside isolated from the roots of Astragalus membranaceus and supplied for research use. It is reported to exert anti-inflammatory effects and is provided as a white to off-white solid for biochemical and pharmacological studies.

  • Flavonoid glycoside isolated from Astragalus membranaceus.
  • Reported anti-inflammatory activity useful for inflammation research.
  • High purity (≈99.9%) suitable for analytical and biological assays.
  • Molecular formula C23H26O10; molecular weight 462.45 g/mol.
  • Available as a 10 mg solid and in additional sizes, including DMSO solution formats.
  • Store at 4°C and protect from light; in solvent: -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-89471 Supplier Medchemexpress LLC Supplier No. HYN247310MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylnissolin-3-O-glucoside is a flavonoid glycoside isolated from the roots of Astragalus membranaceus and supplied for research use. It is reported to exert anti-inflammatory effects and is provided as a white to off-white solid for biochemical and pharmacological studies.

  • Flavonoid glycoside isolated from Astragalus membranaceus.
  • Reported anti-inflammatory activity useful for inflammation research.
  • High purity (≈99.9%) suitable for analytical and biological assays.
  • Molecular formula C23H26O10; molecular weight 462.45 g/mol.
  • Available as a 10 mg solid and in additional sizes, including DMSO solution formats.
  • Store at 4°C and protect from light; in solvent: -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.