Learn More
Medchemexpress LLC HY-N2438 5mg Medchemexpress, Methylophiopogonanone B CAS:74805-91-7 Purity:>98%
SDP
Catalog No. 501970393
Encompass_Preferred
Medchemexpress, HY-N2438 5mg Methylophiopogonanone B CAS:74805-91-7 Methylophiopogonanone B, homoisoflavonoid, is extracted from the root of Ophiopogon japonicas, shows high antioxidant ability. Methylophiopogonanone B increases GTP-Rho and acts via the Rho signaling pathway, inducing cell morphological change via actin cytoskeletal reorganization, including dendrite retraction and stress fiber formation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0393 Supplier Medchemexpress LLC Supplier No. HYN24385MG
Medchemexpress, HY-N2438 5mg Methylophiopogonanone B CAS:74805-91-7 Methylophiopogonanone B, homoisoflavonoid, is extracted from the root of Ophiopogon japonicas, shows high antioxidant ability. Methylophiopogonanone B increases GTP-Rho and acts via the Rho signaling pathway, inducing cell morphological change via actin cytoskeletal reorganization, including dendrite retraction and stress fiber formation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.