Learn More
Cayman Chemical MethylophIopogonon B 10mg
SDP
Catalog No. 502988305
Encompass_Preferred
An isoflavone; increases Rho activity in NHEMs at 1 µM; inhibits LPS-induced NO production in BV-2 microglia (IC50 = 7.8 µM); reduces hydrogen peroxide-induced production of ROS, apoptosis, and decreases in SOD activity in HUVECs at 40 and 50 µM; cytotoxic to SMMC-7721 and HeLa cancer cells (IC50s = 34.6 and 6 UG/ml, respectively)
Catalog No. 50-298-8305 Supplier Cayman Chemical Supplier No. 3019310 MG
An isoflavone; increases Rho activity in NHEMs at 1 µM; inhibits LPS-induced NO production in BV-2 microglia (IC50 = 7.8 µM); reduces hydrogen peroxide-induced production of ROS, apoptosis, and decreases in SOD activity in HUVECs at 40 and 50 µM; cytotoxic to SMMC-7721 and HeLa cancer cells (IC50s = 34.6 and 6 UG/ml, respectively)