Learn More
ATCC Pfizer Pneumococcal; polysaccharide powder; Type 24 (24F) 10mg
Catalog No. 502381166
Encompass
Purified pneumococcal polysaccharide type 24F is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 24F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.
Catalog No. 50-238-1166 Supplier ATCC Supplier No. 552X
Purified pneumococcal polysaccharide type 24F is manufactured by Pfizer, Inc. for small quantity distribution by ATCC. The pneumococcal polysaccharide type 24F can be used to assay for type-specific antibodies, research immune responses, and study immunodeficiencies.