Learn More
U.S. Pharmacopeia Methylparaben, 99-76-3, MFCD00002352, 125mg
Catalog No. NC9738198
Encompass_Preferred
Molecular formula C8H8O3, Molecular weight 152.15, Melting Point 257 - 267.8 °F (125 - 131 °C), Boiling Point 518 - 536 °F (270 - 280 °C) (decomposes), Synonyms: Methyl hydroxybenzoate
Catalog No. NC9738198 Supplier U.S. Pharmacopeia Supplier No. 1432005
Molecular formula C8H8O3, Molecular weight 152.15, Melting Point 257 - 267.8 °F (125 - 131 °C), Boiling Point 518 - 536 °F (270 - 280 °C) (decomposes), Synonyms: Methyl hydroxybenzoate