Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylparaben, FCC, 99-100.5%, Spectrum™ Chemical
SDP

Catalog No. 18604528 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Pail
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-528 500 g Poly Pail
18-604-527 25 g Poly Pail
18-604-529 2.5 kg Poly Pail
18-604-530 12 kg Poly Pail
18-604-531 50 kg Poly Pail
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-528 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1187500GM
Add to Cart
Add to Cart

M1187, 99-76-3, C8H8O3

Spectrum™ Chemical Methylparaben, FCC is used as a food preservative and has the E number E218. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 61-68-7
Molecular Formula C15H15NO2
Molecular Weight (g/mol) 241.29
InChI Key HYYBABOKPJLUIN-UHFFFAOYSA-N
IUPAC Name 2-[(2,3-dimethylphenyl)amino]benzoic acid
SMILES CC1=CC=CC(NC2=CC=CC=C2C(O)=O)=C1C

Specifications

CAS Max % 100%
Ignition Residue 0.05%
Loss on Drying 0.5%
Melting Range 125-128°C
Packaging Poly Pail
Quantity 500 g
Percent Purity 99 to 101%
Grade BP
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.