Learn More
Description
Methylparaben NF / EP can be a white or almost white crystalline powder or as colourless crystals.
Alternative name(s): methyl hydroxybenzoate, methyl 4-hydroxybenzoate, methyl para-hydroxybenzoate, methyl paraben
Specifications
Specifications
| Chemical Name or Material | Methylparaben NF/EP |
| Packaging | Jar, 100mL, White, HDPE, Round, 38-400, TE Pano Neck |
| CAS | 99-76-3 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 152.1482 |
| Quantity | 25 g |
| Percent Purity | 98 to 102 % |
| Grade | EP, NF |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
