Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylparaben Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111016025 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 g
10 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-6025 10 g
11-101-6024 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-6025 Supplier MilliporeSigma Supelco Supplier No. PHR101210G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylparaben is an antifungal and antibacterial agent, widely used as a preservative in cosmetics, pharmaceuticals, food and beverages.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards. Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Pharmaceutical (small molecule)
TRUSTED_SUSTAINABILITY

Specifications

CAS 99-76-3
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP (PhEur) and BP primary standards, where they are available.
Physical Form Neat
Linear Formula HOC6H4 CO2 CH3
MDL Number MFCD00002352
Melting Point 125°C to 128°C
Synonym Methyl 4-hydroxybenzoate; p-Hydroxybenzoic acid methyl ester; Methyl paraben; NIPAGIN
Type Certified Reference Material
Quantity 10 g
Formula Weight 152.15
Grade Certified Reference Material
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.