Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylparaben Sodium, NF, 95-102%, Spectrum™ Chemical
SDP

Catalog No. 18604523 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Poly Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-523 500 g Poly Bottle
18-604-522 125 g Poly Bottle
18-604-524 2.5 kg Poly Bottle
18-604-525 12 kg Poly Bottle
18-604-526 50 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18-604-523 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1277500GM
Only null left
Add to Cart
Add to Cart

M1277, 5026-62-0, C8H7NaO3

Spectrum™ Chemical Methylparaben Sodium, NF is an excipient used as a preservative. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 5026-62-0
Molecular Formula C8H7NaO3
Molecular Weight (g/mol) 174.13
InChI Key PESXGULMKCKJCC-UHFFFAOYSA-M
IUPAC Name sodium 4-(methoxycarbonyl)benzen-1-olate
SMILES [Na+].COC(=O)C1=CC=C([O-])C=C1

Specifications

CAS Max % 100%
Packaging Poly Bottle
Quantity 500 g
Percent Purity 95 to 102%
Grade NF
Physical Form Crystalline Powder or Lumps
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.