Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylphenylvinylsilane, 97%, Thermo Scientific™

Catalog No. AC336680010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC336680010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC336680010 Supplier Thermo Scientific Chemicals Supplier No. 336680010

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Reduction reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17878-39-6
Molecular Formula C9H11Si
Molecular Weight (g/mol) 147.27
MDL Number MFCD00053689
InChI Key MOSXLDGILGBOSZ-UHFFFAOYSA-N
Synonym methylphenylvinylsilane, silane, ethenylmethylphenyl, phenylmethylvinylsilane, benzene, ethenylmethylsilyl, methyl phenyl vinylsilane, ethenyl methyl phenylsilyl, phenyl methyl vinyl silane, methyl phenyl vinyl silane, methyl phenyl vinylsilane #, vinylphenylmethylsilane
PubChem CID 6329252
SMILES C[Si](C=C)C1=CC=CC=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless
Density 0.89g/mL
Infrared Spectrum Authentic
Assay Percent Range 97%
Refractive Index 1.5125 to 1.5145 (20°C, 589nm)
Linear Formula C6H5Si(H)(CH3)CHCH
Quantity 1 g
IUPAC Name ethenyl(methyl)phenylsilyl
Formula Weight 148.28
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Methylphenylvinylsilane
Show More Show Less
Hazard Statement GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 241-830-5
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.