Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylphosphonic dichloride, 98%, Thermo Scientific™

Catalog No. AAA1479006
Change view
Click to view available options
Quantity:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1479006 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAA1479006 Supplier Thermo Scientific Chemicals Supplier No. A1479006

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Methylphosphonic dichloride is used for the preparation of oligonucleotides. It is also used in Suzuki reaction. It serves as a precursor to several chemical weapon agents. Further, it reacts with hydrogen fluoride to get methylphosphonyl difluoride which is used in the production of sarin and soman nerve agents. In addition to this, it undergoes condensation reactions with aminobenzenes and alcohols.

Solubility
Soluble in water.

Notes
Moisture sensitive. Incompatible with strong bases, alcohols and strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Specifications

CAS 676-97-1
Boiling Point 59°C to 60°C (11mmHg)
Flash Point >110°C (230°F)
Odor Acrid
Molecular Formula CH3Cl2OP
Quantity 5g
MDL Number MFCD00002071
UN Number UN2928
Beilstein 1071305
Synonym methylphosphonic dichloride, methylphosphonyl dichloride, methyl phosphonic dichloride, methanephosphonyl chloride, phosphonic dichloride, methyl, methanephosphonic dichloride, methylphosphonic acid dichloride, methylphosphonyl chloride, dichloromethylphosphine oxide, methylphosphonodichloridic acid
InChI Key SCLFRABIDYGTAZ-UHFFFAOYSA-N
SMILES CP(Cl)(Cl)=O
IUPAC Name methylphosphonoyl dichloride
Molecular Weight (g/mol) 132.91
PubChem CID 12671
Formula Weight 132.92
Percent Purity 98%
Chemical Name or Material Methylphosphonic dichloride
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
H330-H314-H318
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement GHS P Statement
P260-P303+P361+P353-P304+P340-P305+P351+P338-P320-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC SOLIDS, CORROSIVE, ORGANIC, N.O.S.
RTECSNumber TA1840000
TSCA TSCA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.