Learn More
Cayman Chemical Methylpiperidino pyrazole, Purity: ≥98%, CAS: 289726-02-9, 50 mg
SDP
Catalog No. 502955017
Encompass_Preferred
A highly selective ERα receptor antagonist (Ki = 5.6 nM; IC50 = 80 nM)
Catalog No. 50-295-5017 Supplier Cayman Chemical Supplier No. 1386350 MG
A highly selective ERα receptor antagonist (Ki = 5.6 nM; IC50 = 80 nM)