Learn More
Medchemexpress LLC HY-B0260 100mg Medchemexpress, Methylprednisolone CAS:83-43-2 Purity:>98%
SDP
Catalog No. 502082430
Encompass_Preferred
Medchemexpress, HY-B0260 100mg Methylprednisolone CAS:83-43-2 Methylprednisolone is a synthetic corticosteroid with anti-inflammatory and immunomodulating properties. Methylprednisolone improve severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2430 Supplier Medchemexpress LLC Supplier No. HYB0260100MG
Medchemexpress, HY-B0260 100mg Methylprednisolone CAS:83-43-2 Methylprednisolone is a synthetic corticosteroid with anti-inflammatory and immunomodulating properties. Methylprednisolone improve severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.