Learn More
Apexbio Technology LLC Methylprednisolone(Synonyms: Medrol, Depo-Medrol, Solu-Medrol, Advantan, Urbason, Metipred, Medrate, Metilprednisolone), 1g, CAS: 83-43-2.
SDP
Catalog No. 5000060175
Encompass_Preferred
Methylprednisolone (CAS 83-43-2) is a synthetic glucocorticoid receptor agonist that exerts anti-inflammatory effects by inhibiting pro-inflammatory cytokines notably TNF- and modulating NF- B signaling pathways In vitro studies demonstrate that methylprednisolone reduces TNF production and enhances IL-10 synthesis in mouse macrophages upon LPS stimulation suppresses chemokine secretion from human peripheral blood mononuclear cells and inhibits acantholysis in skin cultures Animal models indicate that intravenous methylprednisolone administration attenuates inflammation decreases macrophage infiltration and tissue damage post-spinal cord injury Clinically methylprednisolone is investigated for therapeutic use in acute spinal cord injury severe vasculitis and lupus nephritis
Catalog No. 50-000-60175 Supplier Apexbio Technology LLC Supplier No. A42331000
Methylprednisolone (CAS 83-43-2) is a synthetic glucocorticoid receptor agonist that exerts anti-inflammatory effects by inhibiting pro-inflammatory cytokines notably TNF- and modulating NF- B signaling pathways In vitro studies demonstrate that methylprednisolone reduces TNF production and enhances IL-10 synthesis in mouse macrophages upon LPS stimulation suppresses chemokine secretion from human peripheral blood mononuclear cells and inhibits acantholysis in skin cultures Animal models indicate that intravenous methylprednisolone administration attenuates inflammation decreases macrophage infiltration and tissue damage post-spinal cord injury Clinically methylprednisolone is investigated for therapeutic use in acute spinal cord injury severe vasculitis and lupus nephritis