Learn More
U.S. Pharmacopeia Methylprednisolone, 83-43-2, MFCD00010591, 200mg
Catalog No. NC1649729
Encompass
Molecular formula C22-H30-O5, Molecular weight 374.47, Melting point/freezing point 442.4 - 458.6 °F (228 - 237 °C), Synonyms: 6-alpha-Methylprednisolone
Catalog No. NC1649729 Supplier U.S. Pharmacopeia Supplier No. 1435003
Add to Cart
Add to Cart
Molecular formula C22-H30-O5, Molecular weight 374.47, Melting point/freezing point 442.4 - 458.6 °F (228 - 237 °C), Synonyms: 6-alpha-Methylprednisolone