Learn More
Medchemexpress LLC Methylprednisolone | 83-43-2 | 99.78% | 5 G
SDP
Catalog No. 5000406932
Encompass_Preferred
Methylprednisolone is a synthetic corticosteroid with anti-inflammatory and immunomodulating properties. It has been shown to improve severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels. It is commonly used for anti-inflammatory purposes and in the study of various inflammatory conditions.
  • Synthetic corticosteroid with anti-inflammatory and immunomodulating properties
  • Can improve severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels
  • Commonly used for anti-inflammatory purposes
  • Can be used in the study of bronchial inflammation or acute bronchitis caused by arthritis and various respiratory diseases
  • Can improve neurological recovery in acute spinal cord injury
  • Triggers osteoblast death through multiple pathways, inducing femoral head necrosis
Catalog No. 50-004-06932 Supplier Medchemexpress LLC Supplier No. HYB02605G
Methylprednisolone is a synthetic corticosteroid with anti-inflammatory and immunomodulating properties. It has been shown to improve severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels. It is commonly used for anti-inflammatory purposes and in the study of various inflammatory conditions.
  • Synthetic corticosteroid with anti-inflammatory and immunomodulating properties
  • Can improve severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels
  • Commonly used for anti-inflammatory purposes
  • Can be used in the study of bronchial inflammation or acute bronchitis caused by arthritis and various respiratory diseases
  • Can improve neurological recovery in acute spinal cord injury
  • Triggers osteoblast death through multiple pathways, inducing femoral head necrosis