Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methylprednisolone | 83-43-2 | 99.8% | 1 G
SDP

Catalog No. 5000406933
Encompass_Preferred

Methylprednisolone is a synthetic corticosteroid with anti-inflammatory and immunomodulating properties. It improves severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels. This compound is commonly used for anti-inflammatory purposes and can be utilized in the study of bronchial inflammation or acute bronchitis caused by arthritis and various respiratory diseases. It can also improve neurological recovery in acute spinal cord injury.

  • Synthetic corticosteroid
  • Anti-inflammatory and immunomodulating properties
  • Improves severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels
  • Commonly used for anti-inflammatory purposes
  • Can be used in the study of bronchial inflammation or acute bronchitis
  • Can improve neurological recovery in acute spinal cord injury
  • Targets glucocorticoid receptor
  • Molecular formula: C22H30O5
  • Molecular weight: 374.47

Catalog No. 50-004-06933 Supplier Medchemexpress LLC Supplier No. HYB02601G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylprednisolone is a synthetic corticosteroid with anti-inflammatory and immunomodulating properties. It improves severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels. This compound is commonly used for anti-inflammatory purposes and can be utilized in the study of bronchial inflammation or acute bronchitis caused by arthritis and various respiratory diseases. It can also improve neurological recovery in acute spinal cord injury.

  • Synthetic corticosteroid
  • Anti-inflammatory and immunomodulating properties
  • Improves severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels
  • Commonly used for anti-inflammatory purposes
  • Can be used in the study of bronchial inflammation or acute bronchitis
  • Can improve neurological recovery in acute spinal cord injury
  • Targets glucocorticoid receptor
  • Molecular formula: C22H30O5
  • Molecular weight: 374.47

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.