Learn More
TARGETMOL CHEMICALS INC Methylprednisolone Aceponate 2
SDP
Catalog No. 502878551
Encompass_Preferred
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Methylprednisolone Aceponate (SH 440), a non-halogenated corticosteroid, is a potent anti-inflammatory agent. Purity 98%
Catalog No. 50-287-8551 Supplier TARGETMOL CHEMICALS INC Supplier No. T1171L25MG
Also available in 1 mL, 5 mg, 10 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Methylprednisolone Aceponate (SH 440), a non-halogenated corticosteroid, is a potent anti-inflammatory agent. Purity 98%