Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methylprednisolone aceponate | 86401-95-8 | MFCD00866150 | 98.9% | 472.57 g/mol | C27H36O7 | 5 MG
SDP

Catalog No. 5000247463
Encompass_Preferred

Methylprednisolone aceponate is a synthetic glucocorticosteroid provided as a characterized research compound for pharmacology, formulation, and analytical studies. It is supplied as a powder with high purity and defined stability profiles suitable for long-term storage.

  • High purity (≈98.9% by LCMS).
  • Molecular weight 472.57 g/mol.
  • Chemical formula C27H36O7.
  • Stable as powder at -20°C (up to 3 years) and 4°C (up to 2 years).
  • Suitable for pharmacology, topical formulation, and analytical reference work.

Catalog No. 50-002-47463 Supplier Medchemexpress LLC Supplier No. HY1030255MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylprednisolone aceponate is a synthetic glucocorticosteroid provided as a characterized research compound for pharmacology, formulation, and analytical studies. It is supplied as a powder with high purity and defined stability profiles suitable for long-term storage.

  • High purity (≈98.9% by LCMS).
  • Molecular weight 472.57 g/mol.
  • Chemical formula C27H36O7.
  • Stable as powder at -20°C (up to 3 years) and 4°C (up to 2 years).
  • Suitable for pharmacology, topical formulation, and analytical reference work.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.