Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methylprednisolone aceponate | 86401-95-8 | MFCD00866150 | 99.8% | 472.6 g/mol | C27H36O7 | 1 ML
SDP

Catalog No. 5000246100
Encompass_Preferred

Methylprednisolone aceponate is a synthetic glucocorticoid supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for laboratory research. It is intended for in vitro studies of anti-inflammatory and corticosteroid mechanisms and is provided with high purity for reliable assay results.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • High purity (99.8%) suitable for in vitro research.
  • Molecular weight 472.6 g/mol.
  • Chemical formula C27H36O7.
  • Commonly used in anti-inflammatory and corticosteroid research.
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-46100 Supplier Medchemexpress LLC Supplier No. HY10302510MM 1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylprednisolone aceponate is a synthetic glucocorticoid supplied as a ready-to-use 10 mM solution in DMSO (1 mL) for laboratory research. It is intended for in vitro studies of anti-inflammatory and corticosteroid mechanisms and is provided with high purity for reliable assay results.

  • Ready-to-use 10 mM solution in DMSO, 1 mL.
  • High purity (99.8%) suitable for in vitro research.
  • Molecular weight 472.6 g/mol.
  • Chemical formula C27H36O7.
  • Commonly used in anti-inflammatory and corticosteroid research.
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.