Learn More
Selleck Chemical LLC Methylprednisolone Acetate
SDP
Catalog No. 5000003304
Encompass_Preferred
Methylprednisolone (NSC-19987 Medrol) acetate is a synthetic glucocorticoid receptor agonist used to achieve prompt suppression of inflammation Methylprednisolone activates ACE2 and reduces IL-6 levels thus improves severe or critical COVID-19 Methylprednisolone markedly reduces autophagy and apoptosis
Catalog No. 50-000-03304 Supplier Selleck Chemical LLC Supplier No. S59735MG
Methylprednisolone (NSC-19987 Medrol) acetate is a synthetic glucocorticoid receptor agonist used to achieve prompt suppression of inflammation Methylprednisolone activates ACE2 and reduces IL-6 levels thus improves severe or critical COVID-19 Methylprednisolone markedly reduces autophagy and apoptosis