Learn More
Medchemexpress LLC Methylprednisolone-d7 | 83-43-2 | 99.7% | 1 MG
SDP
Catalog No. 5000405592
Encompass_Preferred
Methylprednisolone-d7 is a deuterium-labeled version of Methylprednisolone. Methylprednisolone is a synthetic corticosteroid with anti-inflammatory and immunomodulating properties.
  • Anti-inflammatory properties
  • Immunomodulating properties
  • Can improve severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels
Catalog No. 50-004-05592 Supplier Medchemexpress LLC Supplier No. HYB0260S31MG
Methylprednisolone-d7 is a deuterium-labeled version of Methylprednisolone. Methylprednisolone is a synthetic corticosteroid with anti-inflammatory and immunomodulating properties.
  • Anti-inflammatory properties
  • Immunomodulating properties
  • Can improve severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels