Learn More
Medchemexpress LLC Methylprednisolone (Standard) | 83-43-2 | 99.8% | 25 MG
SDP
Catalog No. 5000450603
Encompass_Preferred
Methylprednisolone is a synthetic corticosteroid possessing anti-inflammatory and immunomodulating properties. It has been shown to improve severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels. Methylprednisolone is commonly utilized for anti-inflammatory purposes, including in the study of bronchial inflammation or acute bronchitis associated with arthritis and various respiratory diseases. It can also enhance neurological recovery in cases of acute spinal cord injury.
  • Synthetic corticosteroid.
  • Exhibits anti-inflammatory properties.
  • Has immunomodulating properties.
  • Can improve severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels.
  • Used in the study of bronchial inflammation or acute bronchitis.
  • May aid in neurological recovery for acute spinal cord injury.
Catalog No. 50-004-50603 Supplier Medchemexpress LLC Supplier No. 25MGHYB0260R
Methylprednisolone is a synthetic corticosteroid possessing anti-inflammatory and immunomodulating properties. It has been shown to improve severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels. Methylprednisolone is commonly utilized for anti-inflammatory purposes, including in the study of bronchial inflammation or acute bronchitis associated with arthritis and various respiratory diseases. It can also enhance neurological recovery in cases of acute spinal cord injury.
  • Synthetic corticosteroid.
  • Exhibits anti-inflammatory properties.
  • Has immunomodulating properties.
  • Can improve severe or critical COVID-19 by activating ACE2 and reducing IL-6 levels.
  • Used in the study of bronchial inflammation or acute bronchitis.
  • May aid in neurological recovery for acute spinal cord injury.