Learn More
Medchemexpress LLC HY-B1900 10mg Medchemexpress, Methylprednisolone succinate CAS:2921-57-5 Purity:>98%
SDP
Catalog No. 501933033
Encompass_Preferred
Medchemexpress, HY-B1900 10mg Methylprednisolone succinate CAS:2921-57-5 Methylprednisolone succinate is a synthetic glucocorticoid and widely used as an anti-inflammatory agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-3033 Supplier Medchemexpress LLC Supplier No. HYB190010MG
Medchemexpress, HY-B1900 10mg Methylprednisolone succinate CAS:2921-57-5 Methylprednisolone succinate is a synthetic glucocorticoid and widely used as an anti-inflammatory agent. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.