Learn More
Gelest Inc [2-3% (mercaptopropyl)methylsiloxane]-dimethylsiloxane copolymer, 120-180 cSt | 102783-03-9 | MFCD00239549 | 100g
Supplier:  Gelest Inc SMS022100G
Encompass
This item has a minimum qty of 14 per supplier requirements.
[2-3% (mercaptopropyl)methylsiloxane]-dimethylsiloxane copolymer, 120-180 cSt | MW: 6,000-8,000 | 102783-03-9 | MFCD00239549 | 100g
Catalog No. NC1391065
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.