Learn More
Cayman Chemical MethylspInazarIn 2.5mg
SDP
Catalog No. 502992182
Encompass_Preferred
A bacterial metabolite and COMT inhibitor (IC50 = 0.8 μg/ml); selective for COMT over tyrosine hydroxylase, DOPA decarboxylase, and dopamine-β-hydroxylase at 100 μg/ml; decreases blood pressure in spontaneously hypertensive rats at 50 mg/kg
Catalog No. 50-299-2182 Supplier Cayman Chemical Supplier No. 298982.5 MG
A bacterial metabolite and COMT inhibitor (IC50 = 0.8 μg/ml); selective for COMT over tyrosine hydroxylase, DOPA decarboxylase, and dopamine-β-hydroxylase at 100 μg/ml; decreases blood pressure in spontaneously hypertensive rats at 50 mg/kg