Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methylsticin | 495-85-2 | 20 MG
SDP

Catalog No. 5000421509
Encompass_Preferred

Methylsticin is a kavalactone isolated from the plant *Piper methysticum*. It exhibits osteoclast formation inhibitory activity, with a molecular weight of 274.27 and a chemical formula of C15H14O5. This compound appears as a white to off-white solid and has demonstrated an IC50 of 0.19 μg/mL in A549 cells for the inhibition of TNF-alpha-induced NF-kappaB expression.

  • Isolated from the plant *Piper methysticum*
  • Exhibits osteoclast formation inhibitory activity
  • White to off-white solid
  • Molecular weight of 274.27
  • Chemical formula of C15H14O5
  • IC50 of 0.19 μg/mL for inhibition of TNF-alpha-induced NF-kappaB expression in A549 cells

Catalog No. 50-004-21509 Supplier Medchemexpress LLC Supplier No. HYN246520MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylsticin is a kavalactone isolated from the plant *Piper methysticum*. It exhibits osteoclast formation inhibitory activity, with a molecular weight of 274.27 and a chemical formula of C15H14O5. This compound appears as a white to off-white solid and has demonstrated an IC50 of 0.19 μg/mL in A549 cells for the inhibition of TNF-alpha-induced NF-kappaB expression.

  • Isolated from the plant *Piper methysticum*
  • Exhibits osteoclast formation inhibitory activity
  • White to off-white solid
  • Molecular weight of 274.27
  • Chemical formula of C15H14O5
  • IC50 of 0.19 μg/mL for inhibition of TNF-alpha-induced NF-kappaB expression in A549 cells

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.