Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Methylsticin | 495-85-2 | MFCD21364778 | 99.8% | 274.27 g/mol | C15H14O5 | 10 MG
SDP

Catalog No. 5000289900
Encompass_Preferred

Methylsticin is a kavalactone isolated from Piper methysticum with reported osteoclast formation inhibitory activity. Supplied as a high-purity analytical standard for research use only, it is available as a solid and as a DMSO stock solution suitable for in vitro and in vivo preparation.

  • High purity: 99.82%.
  • Chemical formula: C15H14O5; molecular weight: 274.27 g/mol.
  • Appearance: white to off-white solid.
  • Solubility: DMSO ~25 mg/mL; in vivo formulation examples provided by supplier.
  • Storage: solid at 4°C protected from light; solutions -80°C up to 6 months, -20°C up to 1 month.
  • Intended for research use only; not for human use.

Catalog No. 50-002-89900 Supplier Medchemexpress LLC Supplier No. HYN246510MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methylsticin is a kavalactone isolated from Piper methysticum with reported osteoclast formation inhibitory activity. Supplied as a high-purity analytical standard for research use only, it is available as a solid and as a DMSO stock solution suitable for in vitro and in vivo preparation.

  • High purity: 99.82%.
  • Chemical formula: C15H14O5; molecular weight: 274.27 g/mol.
  • Appearance: white to off-white solid.
  • Solubility: DMSO ~25 mg/mL; in vivo formulation examples provided by supplier.
  • Storage: solid at 4°C protected from light; solutions -80°C up to 6 months, -20°C up to 1 month.
  • Intended for research use only; not for human use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.