Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methylsulfamic Acid 98.0+%, TCI America™
SDP

Catalog No. M10835G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M10835G 5 g
M108325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M10835G Supplier TCI America Supplier No. M10835G
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4112-03-2
Molecular Formula CH5NO3S
Molecular Weight (g/mol) 111.12
MDL Number MFCD00010870
InChI Key MYMDOKBFMTVEGE-UHFFFAOYSA-N
PubChem CID 244258
IUPAC Name methylsulfamic acid
SMILES CNS(=O)(=O)O

Specifications

Color White-Yellow
Quantity 5 g
UN Number 1759
Formula Weight 111.12
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Methylsulfamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.